Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium nitride, Aerosol Refractory Paint, ≥99%, Thermo Scientific™

Catalog No. AA40390CLC
Change view
Click to view available options
Quantity:
13 oz.
5 x 13 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40390CLC 13 oz.
AA403900CL 5 x 13 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40390CLC Supplier Thermo Scientific Chemicals Supplier No. 40390CLC

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.87
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
PubChem CID 93091
IUPAC Name titanioamine
SMILES N#[Ti]

Specifications

Color Black-Brown
Quantity 13 oz.
UN Number UN1950
Solubility Information Soluble in aqua regain. Partly soluble in water.
Percent Purity ≥99%
Odor Alcohol-like
Packaging Aerosol Spray Can
Chemical Name or Material Titanium nitride, Aerosol Refractory Paint
Hazard Category H224-H319-H336-X
Hazard Statement GHS H Statement
H220-H319-H336
Extremely flammable gas.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P211-P235-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.