Learn More
Strem, An Ascensus Company CAS# 7440-32-6. 500g. Titanium sponge (99.5%). MFCD00011264
SDP
Catalog No. 5090114467
Encompass
CAS# 7440-32-6. 500g. Titanium sponge (99.5%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: random pieces Strem# 93-2243. http://www.strem.com/catalog/v/93-2243/
Catalog No. 50-901-14467 Supplier Strem, An Ascensus Company Supplier No. 932243500G
CAS# 7440-32-6. 500g. Titanium sponge (99.5%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: random pieces Strem# 93-2243. http://www.strem.com/catalog/v/93-2243/