Learn More
Takara Bio Titanium Taq DNA Polymerase 100 Rxns
SDP
Catalog No. NC9806143
Encompass
Titanium Taq DNA Polymerase is specially engineered for higher robustness and sensitivity than wild-type Taq, and contains TaqStart Antibody for hot-start PCR. The polymerase mix is suitable for use in all PCR applications. A separate tube of optimized PCR buffer (Mg2+ plus) is supplied as well. Enough enzyme and buffer are supplied for PCR reactions of 50 l each. 100 Rxns
Catalog No. NC9806143 Supplier Takara Bio Supplier No. 639208
Titanium Taq DNA Polymerase is specially engineered for higher robustness and sensitivity than wild-type Taq, and contains TaqStart Antibody for hot-start PCR. The polymerase mix is suitable for use in all PCR applications. A separate tube of optimized PCR buffer (Mg2+ plus) is supplied as well. Enough enzyme and buffer are supplied for PCR reactions of 50 l each. 100 Rxns