Learn More
Takara Bio TITANIUM TAQ POLYMRS 500X
SDP
Catalog No. NC9793498
Encompass
TITANIUM Taq DNA Polymerase,500 rxns, Higher yields for more reproducible results, High sensitivity-ideal for limited starting material, Built-in hot start-ideal for multiplex PCR, No Mg2 + optimization or 10-min. activation step required
Catalog No. NC9793498 Supplier Takara Bio Supplier No. 639209
TITANIUM Taq DNA Polymerase,500 rxns, Higher yields for more reproducible results, High sensitivity-ideal for limited starting material, Built-in hot start-ideal for multiplex PCR, No Mg2 + optimization or 10-min. activation step required