Learn More
Pfaltz & Bauer Titanium trichloride 20% solution in 2N HCl, 7705-07-9, MFCD00011266, 500 mL
Catalog No. NC0750648
Encompass
TITANIUM(III) CHLORIDE, Molecular Formula: Cl3 Ti, Molecular Weight: 154.226, Boiling Point: 100, Density: 1.192, DOT Name: Corrosive Liquid, Acidic, Inorganic, N.O.S.
Catalog No. NC0750648 Supplier Pfaltz & Bauer Supplier No. T13605500ML
TITANIUM(III) CHLORIDE, Molecular Formula: Cl3 Ti, Molecular Weight: 154.226, Boiling Point: 100, Density: 1.192, DOT Name: Corrosive Liquid, Acidic, Inorganic, N.O.S.