Learn More
Sigma Aldrich Fine Chemicals Biosciences Titanium tweezers 6 piece1EA
Catalog No. 502901512
Encompass
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight high strength non-magnetic material. Lightweight (high strength to weight ratio)Zero magnetic retentivelyTemperature resistant to 800 fChemical and corrosion resistantNon-solder adhesiveBio-compatible (maintain cell integrity no inflammatory response)Typical application handling of sensitive components in cleaning/chemical processes at high temperatures histology biology forensics electron microscopy etc.
Catalog No. 50-290-1512 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6802731EA
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight high strength non-magnetic material. Lightweight (high strength to weight ratio)Zero magnetic retentivelyTemperature resistant to 800 fChemical and corrosion resistantNon-solder adhesiveBio-compatible (maintain cell integrity no inflammatory response)Typical application handling of sensitive components in cleaning/chemical processes at high temperatures histology biology forensics electron microscopy etc.