Learn More
Strem, An Ascensus Company CAS# 7440-32-6. 10m. Titanium wire (99.7%). MFCD00011264
SDP
Catalog No. 5090114479
Encompass
CAS# 7440-32-6. 10m. Titanium wire (99.7%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: 0.5 mm dia. Strem# 93-2253. http://www.strem.com/catalog/v/93-2253/
Catalog No. 50-901-14479 Supplier Strem, An Ascensus Company Supplier No. 93225310MT
CAS# 7440-32-6. 10m. Titanium wire (99.7%). MFCD00011264. Molecular Weight: 47.90. Molecular Formula: Ti. Color/form: 0.5 mm dia. Strem# 93-2253. http://www.strem.com/catalog/v/93-2253/