Learn More
Sigma Aldrich Fine Chemicals Biosciences TitaniumIV isopropoxide 97
Catalog No. 502899111
Encompass
Titanium(IV) isopropoxide is a titanium-based coordination compound commonly used in the asymmetric Sharpless epoxidation reaction of allylic alcohols. It is also used as a catalyst in Kulinkovich reaction for the synthesis of cyclopropanes.
Catalog No. 50-289-9111 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2052734X25ML
Titanium(IV) isopropoxide is a titanium-based coordination compound commonly used in the asymmetric Sharpless epoxidation reaction of allylic alcohols. It is also used as a catalyst in Kulinkovich reaction for the synthesis of cyclopropanes.