Learn More
GL Sciences Inc Titansphere TiO, 5um | 500mg
Catalog No. NC1701094
Encompass
Spherically shaped porous particle. Provide a high recovery rate even with small amount of sample. Since it can specifically adsorb phosphate sites, it can be applicable to the purification of phosphorylated peptides, phosphorylated sugars, glyphosate, etc.
Catalog No. NC1701094 Supplier GL Sciences Inc Supplier No. 502075000
Spherically shaped porous particle. Provide a high recovery rate even with small amount of sample. Since it can specifically adsorb phosphate sites, it can be applicable to the purification of phosphorylated peptides, phosphorylated sugars, glyphosate, etc.