Learn More
Expression Systems Baculovirus Titering Kit (includes gp64-PE antibody and Baculovirus control)
SDP
Catalog No. NC9612746
Encompass_Preferred
Expression Systems’ Baculovirus Titering Kit facilitates accurate determination of infectious baculovirus titers in less than 24 hours. The assay is based on detection of the baculovirus gp64 fusion protein which is expressed on the surface of infected insect cells within six hours of infection. The kit contains gp64-PE antibody and a baculovirus control of known titer.
• Reproducible and accurate titer results within 24 hours
• Measures infectious virus
• Suspension culture methodology simulates actual production conditions
• Works with any flow cytometer capable of reading samples from 96-well plates
• Flow cytometric data collection removes the operator error of counting plaques
• gp64-PE antibody can be used alone to monitor infection kinetics of baculovirus in insect cell culture
Catalog No. NC9612746 Supplier Expression Systems Supplier No. 97101
Expression Systems’ Baculovirus Titering Kit facilitates accurate determination of infectious baculovirus titers in less than 24 hours. The assay is based on detection of the baculovirus gp64 fusion protein which is expressed on the surface of infected insect cells within six hours of infection. The kit contains gp64-PE antibody and a baculovirus control of known titer.
• Reproducible and accurate titer results within 24 hours
• Measures infectious virus
• Suspension culture methodology simulates actual production conditions
• Works with any flow cytometer capable of reading samples from 96-well plates
• Flow cytometric data collection removes the operator error of counting plaques
• gp64-PE antibody can be used alone to monitor infection kinetics of baculovirus in insect cell culture