Learn More
Sigma Aldrich Fine Chemicals Biosciences Titer Tops sealing film for microplates, 100/pk
Catalog No. NC1452339
Encompass
Durable and thick (4.4 mil) yet easily pierced with a pipette tip. Prevents well-to-well contamination, sample spillage and evaporation. Polyethylene film that provides excellent adhesion to plastic microplates. Non-cytotoxic, sterile, solvent resistant and waterproof.
Catalog No. NC1452339 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6886301PAK
Durable and thick (4.4 mil) yet easily pierced with a pipette tip. Prevents well-to-well contamination, sample spillage and evaporation. Polyethylene film that provides excellent adhesion to plastic microplates. Non-cytotoxic, sterile, solvent resistant and waterproof.