Learn More
USA Scientific Inc Titer-Tops film, pierceable, sterile, 50/bx
Catalog No. NC9504238
Encompass
Titer-Tops polyethylene film, pierceable, 4.4 mil thick, -40C to +100C, sterile, 50/box
Catalog No. NC9504238 Supplier USA Scientific Inc Supplier No. 91886775
Titer-Tops polyethylene film, pierceable, 4.4 mil thick, -40C to +100C, sterile, 50/box