Learn More
Electron Microscopy Sciences Titer-Tops, 6" X 3.25"
SDP
Catalog No. 50364989
Encompass_Preferred
Titer-Tops prevents sample evaporation, well to well contamination, external contamination and sample spillage and are made from 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive. Each pre-cut rectangular sheet is 6"x3.25" with a paper liner. The center section of the liner measures 5"x3.25" and once it is peeled off it leaves two hand-grip extension tabs on the two ends allowing for easy application of the Titer-Top to the plate.
Titer-tops are chemically resistant and thermally stable and the acrylic adhesive is waterproof, solvent resistant, and compatible with all plastic microtiter plates. They will stay in place even when exposed to 100% humidity and temperatures ranging from -40 degree C to 100 degree C. They are easily pierced with a micropipette tip to provide access to sample wells. Gamma sterile.
Catalog No. 50-364-989 Supplier Electron Microscopy Sciences Supplier No. 7053805
Titer-Tops prevents sample evaporation, well to well contamination, external contamination and sample spillage and are made from 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive. Each pre-cut rectangular sheet is 6"x3.25" with a paper liner. The center section of the liner measures 5"x3.25" and once it is peeled off it leaves two hand-grip extension tabs on the two ends allowing for easy application of the Titer-Top to the plate.
Titer-tops are chemically resistant and thermally stable and the acrylic adhesive is waterproof, solvent resistant, and compatible with all plastic microtiter plates. They will stay in place even when exposed to 100% humidity and temperatures ranging from -40 degree C to 100 degree C. They are easily pierced with a micropipette tip to provide access to sample wells. Gamma sterile.