Learn More
Andwin Scientific TITER-TOPS NON-CYTOTOXIC GAM
SDP
Catalog No. 50821033
Encompass_Preferred
The Titer-Tops film is a sealing film with polyethylene base, acrylic adhesive. It features thickness of 112 m and diameter of 152.4 x 82.6 mm. It is UV transparent to 250 nm. The film can be pierced with a pipet tip. It is also sterilized using gamma rays and is non-cytotoxic.
Catalog No. 50-821-033 Supplier Andwin Scientific Supplier No. 91886775
The Titer-Tops film is a sealing film with polyethylene base, acrylic adhesive. It features thickness of 112 m and diameter of 152.4 x 82.6 mm. It is UV transparent to 250 nm. The film can be pierced with a pipet tip. It is also sterilized using gamma rays and is non-cytotoxic.