Learn More
Sigma Aldrich Fine Chemicals Biosciences TiterMaxR Classic Adjuvant
Catalog No. 502900696
Encompass
The effectiveness of an oil emulsion vaccine greatly depends on the form and stability of the emulsion. TiterMax Classic is a water-in-oil formulation with up to 90% water and a powerful adjuvant used in many protocols. TiterMax Classic Adjuvant contains three essential ingredients a block copolymer CRL89-41 squalene (a metabolizable oil) and a unique microparticulate stabilizer. The emulsion is less viscous and suitable for injection through small needles.
Catalog No. 50-290-0696 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H4397.5ML
The effectiveness of an oil emulsion vaccine greatly depends on the form and stability of the emulsion. TiterMax Classic is a water-in-oil formulation with up to 90% water and a powerful adjuvant used in many protocols. TiterMax Classic Adjuvant contains three essential ingredients a block copolymer CRL89-41 squalene (a metabolizable oil) and a unique microparticulate stabilizer. The emulsion is less viscous and suitable for injection through small needles.