Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips

Catalog No. p-4605385 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Sterility:
Sterile
Format:
Rack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
ICN77944H2 Sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN77944H2 Supplier MP Biomedicals™ Supplier No. 0977944H2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrophobic tips with very low coefficient of friction

Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette

 

  • Made from virgin polypropylene material
  • Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
  • 96 tips per rack
  • Volume: 1500μL

 

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1500 μL
Sterility Sterile
Color Natural
Format Rack
Material Polypropylene
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.