Learn More
Takara Bio TITIAN DNA AMPLFTN 300R/A
SDP
Catalog No. NC9522029
Encompass
DNA Amplification Kit; TITANIUM; Use w/New GeneChip Mapping 500K Assay; Contains TITANIUM Taq Polymerase; 10X TITANIUM Taq Buffer; GC-Melt Reagent; dNTPs to run 300 PCR rxn in Support of the Affymetrix GeneChip Mapping 500K Assay
Catalog No. NC9522029 Supplier Takara Bio Supplier No. 639240
DNA Amplification Kit; TITANIUM; Use w/New GeneChip Mapping 500K Assay; Contains TITANIUM Taq Polymerase; 10X TITANIUM Taq Buffer; GC-Melt Reagent; dNTPs to run 300 PCR rxn in Support of the Affymetrix GeneChip Mapping 500K Assay