Learn More
Enzo Life Sciences Titin monoclonal antibody (F146.9B9) (5ml)
SDP
Supplier:  Enzo Life Sciences ALXBC3010SL005
Encompass_Preferred
Clone: F146.9B9. Isotype: IgG1. Immunogen: Purified titin from human skeletal muscle. Species reactivity: Human, Mouse Rabbit. Applications: IHC (FS), WB. Formulation: 1ml/5ml of sterile filtered supernatant containing 0.1% sodium azide.
Catalog No. 50-201-0173