Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Automation Setup Kit

Catalog No. 502557496
Encompass_Preferred
Change view
Click to view available options
Quantity:
6 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-7496 6 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-7496 Supplier Invitrogen™ Supplier No. EPXSETUP384
Add to Cart
Add to Cart

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol.

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol, such as liquid handling and plate washing, before running an actual assay with valuable sample. The user guide describes how to adjust and optimize an automated magnetic plate washer as needed during the ProcartaPlex 384-well assay workflow in order to ensure optimal performance and avoid bead loss.

Each kit provides six optimization tests for automated magnetic plate washing.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Basic and Setup Kits
Quantity 6 Tests
For Use With (Equipment) FLEXMAP 3 D™ INTELLIFLEX™
Product Line ProcartaPlex
Content And Storage

• Automation Setup Beads (1X)
• 1 x 200 mL Wash Buffer (10X)
• 1 Bottle Reading Buffer (1X)
• Adhesive film
• ProcartaPlex 384-well flat bottom plate

Store at 2°C to 8°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.