Learn More
McKesson General Medical Titmus V4 Vision Screener, Stereoscopic Instrument, Comprehensive Visual Testing, 2 Years Warranty
Catalog No. NC3351845
Encompass_Preferred
The Titmus V4 Vision Screener is a compact and portable instrument designed for efficient visual performance assessment in clinical and occupational settings, catering to both adults and children. It accurately measures visual acuity, depth perception, and color perception, enhancing test reliability with features like an ergonomic design and fluorescent light source that mimics daylight. This versatility differentiates it from other screeners, making it an ideal choice for comprehensive vision testing.
This item is not returnable. View return policy
Catalog No. NC3351845 Supplier McKesson General Medical Supplier No. 747661
Add to Cart
Add to Cart
Request a Quote
The Titmus V4 Vision Screener is a compact and portable instrument designed for efficient visual performance assessment in clinical and occupational settings, catering to both adults and children. It accurately measures visual acuity, depth perception, and color perception, enhancing test reliability with features like an ergonomic design and fluorescent light source that mimics daylight. This versatility differentiates it from other screeners, making it an ideal choice for comprehensive vision testing.