Learn More
Andwin Scientific TITRA VESSEL METTLER NO101974
SDP
Catalog No. NC9485544
Encompass_Preferred
The 100 mL Mettler Toledo titration beaker provide accuracy with graduation marks. The beaker has a sturdy design and a disposability feature. It is made of standard polypropylene that is efficient for light-sensitive samples. This product is sold in a set of 1400 beakers.
Catalog No. NC9485544 Supplier Andwin Scientific Supplier No. 101974
The 100 mL Mettler Toledo titration beaker provide accuracy with graduation marks. The beaker has a sturdy design and a disposability feature. It is made of standard polypropylene that is efficient for light-sensitive samples. This product is sold in a set of 1400 beakers.