Learn More
Hach Company Titralab AS1000 Sample Changer
Catalog No. 5000207746 Shop All Hach Company Products
Encompass
Titralab AS1000 Series Sample Changer-30 Positions-50 mL BeakersThis application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and two (2) high precision glass 25 mL syringes with syringe rings The Titralab AS1000 series is an automatic sample changer used in analytical laboratories in connection with AT1000 Series titrators The AS1000 is available in four configurations of different tray capacity that holds the sample beakers AS1000 Sample Changer contains Lifting module-Rotating module with carousel tray-support base with magnetic stirrer-probe tubes holder-RS-232 cable with USB/Serial adapter to connect AS1000 to PC-Power supply-set of magnetic stir bars-set of PP beakers TM1000 PC-Software with ethernet cables to connect AT1000 to local network or PC The AT1000 Instrument and Reagents are sold separately
Catalog No. 50-002-07746 Supplier Hach Company Supplier No. AS1000.97.30050
Add to Cart
Add to Cart
Request a Quote
Titralab AS1000 Series Sample Changer-30 Positions-50 mL BeakersThis application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and two (2) high precision glass 25 mL syringes with syringe rings The Titralab AS1000 series is an automatic sample changer used in analytical laboratories in connection with AT1000 Series titrators The AS1000 is available in four configurations of different tray capacity that holds the sample beakers AS1000 Sample Changer contains Lifting module-Rotating module with carousel tray-support base with magnetic stirrer-probe tubes holder-RS-232 cable with USB/Serial adapter to connect AS1000 to PC-Power supply-set of magnetic stir bars-set of PP beakers TM1000 PC-Software with ethernet cables to connect AT1000 to local network or PC The AT1000 Instrument and Reagents are sold separately