Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titrant 5, MilliporeSigma™

Catalog No. M1880100545 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880100545 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1880100545 Supplier MilliporeSigma Supplier No. 1.88010.0545
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Titrant 5 titrant for volumetric Karl Fischer titration with two component reagents H2O Aquastar™.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.85 g/cm3 (at 20°C)
Quantity 500 mL
Flash Point < 21°C
Packaging Glass Bottle
Grade Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.