Learn More
BURKLE INC Titrating burette automatic, borosil. glass, 0.51 oz. (15 ml)
Catalog No. 5000025566
Encompass_Preferred
Burette with Schellbach stripes type Schilling / Bürkle with original Bürkle dosing armature.
Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet. Graduation according to Category B (Tolerances according to DIN EN ISO 385).
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass
Catalog No. 50-000-25566 Supplier BURKLE INC Supplier No. 96951015
Burette with Schellbach stripes type Schilling / Bürkle with original Bürkle dosing armature.
Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet. Graduation according to Category B (Tolerances according to DIN EN ISO 385).
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass