Learn More
BURKLE INC Titrating burette with shatter protection 0.51 oz. (15 ml)
Catalog No. 5000025890
Encompass_Preferred
The titrating burette with splinter protection is a fast acting burette with Schellbach stripes based on Dr. Schilling's design with automatic zero point extraction and Bürkle dosing armature.
The glass burette has a plastic sheathing which increases resistance to breaking and protects against splinters. The possibility of the burette breaking is much lower than with a conventional burette, and if it does break, there will be no dangerous splinters.
The Bürkle dosing armature allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet.
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass
Catalog No. 50-000-25890 Supplier BURKLE INC Supplier No. 96953015
The titrating burette with splinter protection is a fast acting burette with Schellbach stripes based on Dr. Schilling's design with automatic zero point extraction and Bürkle dosing armature.
The glass burette has a plastic sheathing which increases resistance to breaking and protects against splinters. The possibility of the burette breaking is much lower than with a conventional burette, and if it does break, there will be no dangerous splinters.
The Bürkle dosing armature allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet.
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass