Learn More
BURKLE INC Titrating burette automatic, borosil. glass, 0.85 oz. (25 ml)
Catalog No. NC2187891
Encompass_Preferred
Burette with Schellbach stripes type Schilling / Bürkle with original Bürkle dosing armature.
Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet. Graduation according to Category B (Tolerances according to DIN EN ISO 385).
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass
This item is not returnable. View return policy
Catalog No. NC2187891 Supplier BURKLE INC Supplier No. 96951025
Burette with Schellbach stripes type Schilling / Bürkle with original Bürkle dosing armature.
Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet. Graduation according to Category B (Tolerances according to DIN EN ISO 385).
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass