Learn More
Hach Company Titration Application Pack Bev
Catalog No. 5000207351 Shop All Hach Company Products
Encompass
Titration Application Pack Beverage pH-Total Acidity in Beer-Kombucha-Soft Drink/Juice/Energy Drink Bottlers-Single 25 mLApplication Pack for the determination of pH Total Acidity in Beer Kombucha Soft Drink/Juice/Energy Drink Bottlers Suited for Titralab AT1000 Series Automatic Titrator Models AT1102/AT1112/AT1122/AT1222 This application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and one (1) high precision glass 25 mL syringe with syringe ring The AT1000 Instrument and Reagents are sold separately
Catalog No. 50-002-07351 Supplier Hach Company Supplier No. AP0025.AT1102
Titration Application Pack Beverage pH-Total Acidity in Beer-Kombucha-Soft Drink/Juice/Energy Drink Bottlers-Single 25 mLApplication Pack for the determination of pH Total Acidity in Beer Kombucha Soft Drink/Juice/Energy Drink Bottlers Suited for Titralab AT1000 Series Automatic Titrator Models AT1102/AT1112/AT1122/AT1222 This application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and one (1) high precision glass 25 mL syringe with syringe ring The AT1000 Instrument and Reagents are sold separately