Learn More
Hach Company Titration Application Pack Bev
Catalog No. 5000207680 Shop All Hach Company Products
Encompass
Titration Application Pack Beverage pH-Total Acidity in Soft Drink/Juice/ Energy Drink Bottlers-Double 25 mLApplication Pack for the determination of pH Total Acidity in Soft Drink/Juice/ Energy Drink Bottlers Double 25 mL Suited for Titralab AT1000 Series Automatic Titrator Model AT1222 This application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and two (2) high precision glass 25 mL syringes with syringe rings The AT1000 Instrument and Reagents are sold separately
Catalog No. 50-002-07680 Supplier Hach Company Supplier No. AP0026.AT1222
Titration Application Pack Beverage pH-Total Acidity in Soft Drink/Juice/ Energy Drink Bottlers-Double 25 mLApplication Pack for the determination of pH Total Acidity in Soft Drink/Juice/ Energy Drink Bottlers Double 25 mL Suited for Titralab AT1000 Series Automatic Titrator Model AT1222 This application pack contains a pH probe-USB key with pre-programmed applications-polypropylene beakers-magnetic stirring bars-bottle stoppers-conical adapter and two (2) high precision glass 25 mL syringes with syringe rings The AT1000 Instrument and Reagents are sold separately