Learn More
Mettler Toledo Titration Arm GL80
Catalog No. 01672601 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Titration Arm
Catalog No. Type
01-672-601 Titration Arm
1 options
Catalog No. 01-672-601 Supplier Mettler Toledo Supplier No. 30988536
Add to Cart
Add to Cart
A modular accessory designed for (KF) titrators especially for the EVA series, it is part of a broader accessory kit often included in a cell kit
  • The titration arm serves as the structural / mechanical component that holds and supports the titration vessel (cell), electrodes, and other components when performing a KF titration.
  • With the arm in place, the titration cell becomes stable and ready for use enabling either coulometric (electro-chemical generation of titrant) or volumetric (burette-based titrant addition) Karl-Fischer titration depending on your setup.
  • It helps ensure correct orientation, proper sealing and safe handling which is especially critical in KF titrations because moisture ingress or leaks can compromise the accuracy.
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Arm