Learn More
Leica Biosystems TITRATION KIT
Catalog No. NC0209406
Encompass
The BOND Titration Kit contains BOND Titration Container Inserts and BOND Titration Containers.The kit lets users optimize primary antibody con-centrates on the BOND system.The kits can be re-used for different antibodies and are designed with minimal dead volume to preserve reagent.
Catalog No. NC0209406 Supplier Leica Biosystems Supplier No. OPT9049
The BOND Titration Kit contains BOND Titration Container Inserts and BOND Titration Containers.The kit lets users optimize primary antibody con-centrates on the BOND system.The kits can be re-used for different antibodies and are designed with minimal dead volume to preserve reagent.