Learn More
Mettler Toledo Titration Kit KFC
Catalog No. 01672597 Shop All METTLER TOLEDO Products
Catalog No. 01-672-597 Supplier Mettler Toledo Supplier No. 30988520
Add to Cart
Add to Cart
Provides all essential components needed to set up a Karl Fischer cell for moisture/water content determination, enabling a KF-capable titrator to operate properly
  • The Titration kit KFC is a complete cell-kit (accessory set) designed for coulometric Karl Fischer titration (KF), sold by Mettler Toledo.
  • The Titration kit KFC includes a titration arm, a vessel, and a complete set of parts for the EVA Coulometric Karl Fischer cell.
  • Compatible with C1X, C1D, C3X, and C3D models.