Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titration Solvent Blend D4739, Solstice

Catalog No. 60090635 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-635 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-635 Supplier Solstice Supplier No. WP216219P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend, Total Base Number Titration Solvent (ASTM D 4739)

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-66-3, 108-88-3, 67-63-0, 7732-18-5
CAS Max % 47.40%, 27.60%, 24.90%, 0.10%
Quantity 19 L
Flash Point 18.3°C
Packaging Winpak
Physical Form Liquid
Purity Grade Notes Custom Labready™ blends
Boiling Point 75.56°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.