Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titration Solvent Blend D664, Solstice

Catalog No. 60048617 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-617 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-617 Supplier Solstice Supplier No. 21484X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend, Total Acid Number Titration Solvent (ASTM D 664)

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-88-3, 67-63-0, 7732-18-5
Boiling Point 81.1°C
CAS Max % 52.40%, 47.00%, 0.60%
Purity Grade Notes Other catalog blends
Packaging Glass Bottle
Quantity 4 x 4 L
Flash Point -5°C
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.