Learn More
Titration Solvent with Chloroform, ASTM D 4739, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Titration Solvent with Chloroform |
| Color | Colorless |
| CAS | 67-66-3, 108-88-3, 67-63-0, 7732-18-5 |
| CAS Min % | 46.0, 26.85, 24.21, 0.93 |
| CAS Max % | 48.0, 27.95, 25.2, 0.97 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Quantity | 4 L |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.