Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titration Solvent, for Neutralization Number of Petroleum Products, Ricca Chemical
SDP

Catalog No. R86200001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R86200001C 1 L
8620-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R86200001C Supplier Ricca Chemical Company Supplier No. 862032
Add to Cart
Add to Cart

Note: May not be shipped via UPS Air Services

Specifications

Chemical Name or Material Titration Solvent
Name Note For Neutralization Number of Petroleum Products
CAS 108-88-3, 67-63-0, 7732-18-5
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.