Learn More
Metrohm USA TITRATION STAND MODEL 803TI
SDP
Supplier:  Metrohm USA 28030010
Encompass
Compact titration stand with magnetic stirrer and built-in pump for solvent addition and aspiration of the titration vessel contents. Suitable for Karl Fischer titrations with Titrandos. MSB connection. Including comprehensive accessories, solvent bottle and waste bottle.
Catalog No. NC0269964