Learn More
Metrohm USA TITRATION VESSEL
SDP
Supplier:  Metrohm USA 61464320
Encompass
KF titration vessel / 80-250 mL / coulometric. Height: 114 mm. Material: borosilicate glass 3.3.
Catalog No. NC0119991