Learn More
GFS Chemicals Inc TITRATION SOLVENT, ASTM D664 | 4 X 4 L | GFS CHEMICALS, INC. Item #: 74912 | Bulk #: 1281
SDP
Catalog No. NC9613474
Encompass_Preferred
TITRATION SOLVENT, ASTM D664 | Chemical Formula : | CAS # : 0 | Specifications : Appearance - Clear, colorless liquid, Density (g/ml) at 25 C 0.817 - 0.827, H2O (KF) 0.4 - 0.6 %, made per ASTM D664, section 7.18, Suitability, Titrable acid or base 0.0001 meq/g
Catalog No. NC9613474 Supplier GFS Chemicals Inc Supplier No. 74912
TITRATION SOLVENT, ASTM D664 | Chemical Formula : | CAS # : 0 | Specifications : Appearance - Clear, colorless liquid, Density (g/ml) at 25 C 0.817 - 0.827, H2O (KF) 0.4 - 0.6 %, made per ASTM D664, section 7.18, Suitability, Titrable acid or base 0.0001 meq/g