Learn More
Mettler Toledo Titrator Karl Fischer EVA C1 D
Catalog No. 01672595 Shop All METTLER TOLEDO Products
Catalog No. 01-672-595 Supplier Mettler Toledo Supplier No. 31028041
Earn Up to $2,000 in Accessories with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
A cutting-edge solution designed for precise and efficient trace water determination across diverse sample types
  • The EVA C1D Coulometric Karl Fischer Titrator by METTLER TOLEDO.
  • System includes the main unit, terminal, generator electrode with a diaphragm, titration vessel kit, and an automated solvent exchange system for safer and more efficient workflows.
  • Innovative cell design enables easy cleaning and reduces cross-contamination risk.
  • Robust Swiss-made system ensures reliability and longevity even under heavy use.