Learn More
Mettler Toledo Titrator Karl Fischer EVA C1 X
Catalog No. 01672593 Shop All METTLER TOLEDO Products
Catalog No. 01-672-593 Supplier Mettler Toledo Supplier No. 30988510
Earn Up to $2,000 in Accessories with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
A cutting-edge (EVA C1 X) solution designed for precise and efficient trace water determination across diverse sample types
  • The EVA C1X Coulometric Karl Fischer Titrator by METTLER TOLEDO.
  • The system includes the main unit, terminal, diaphragmless generator electrode, titration vessel kit, and an automated solvent exchange system for safer and more efficient workflows.
  • Featuring an innovative cell design for easy cleaning and reduced cross-contamination risk, this robust Swiss-made system ensures reliability and longevity even under heavy use.
  • Safety is paramount with the dPumpKF automatic solvent exchange functions that minimize chemical exposure.
  • An extended terminal cable allows convenient operation outside fume hoods. Digital workflows are streamlined via LabX™ Software, supporting compliance (including 21 CFR Part 11), centralized data management, and effortless process control.
  • For increased productivity, the EVA C1 can be paired with the InMotion KF Six oven autosampler, automating throughput and challenging sample analysis.
  • Experience fast, reliable, and automated water determination with the EVA C1X an advanced KF titration system for modern laboratories.