Learn More
Mettler Toledo Titrator Karl Fischer EVA C3 D
Catalog No. 01672596 Shop All METTLER TOLEDO Products
Catalog No. 01-672-596 Supplier Mettler Toledo Supplier No. 31028043
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
EVA C3 D an advanced instrument crafted for precise trace water determination in various sample types
  • The EVA C3D Coulometric Karl Fischer Titrator by METTLER TOLEDO.
  • System includes the main unit, terminal, generator electrode with diaphragm, titration vessel kit, and an automated solvent exchange system for safer and more efficient workflows.
  • Innovative cell design simplifies cleaning and reduces cross-contamination risks.
  • Integrates seamlessly with balances for automatic, error-free sample weight transfer to enhance accuracy and efficiency.
  • Designed for safety and convenience with automatic solvent exchange using the dPumpKF and an extendable terminal cable for remote operation outside fume hoods.
  • Expanded automation capabilities through compatibility with InMotion KF Six, Flex, and Pro oven autosamplers for high-throughput and challenging sample analysis.
  • Robust titrator supports comprehensive user management with customizable access rights for secure operation.
  • Integration with LabX™ Software streamlines compliance with 21 CFR Part 11, supports ALCOA++ principles, and enables centralized data management and process control.
  • Provides fast, reliable, and automated water determination, making it an ultimate KF titration system for modern laboratories.