Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo Titrator Karl Fischer EVA C3 X

Catalog No. 01672594 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Depth (English):
7 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
01-672-594 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-594 Supplier Mettler Toledo Supplier No. 30988512
Earn Up to $2,000 in Accessories with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

An advanced Titrator EVA C3 X crafted for precise trace water determination in various sample types

  • The EVA C3X Coulometric Karl Fischer Titrator by METTLER TOLEDO is an advanced instrument designed for precise trace water determination in various sample types.
  • System includes the main unit, terminal, diaphragmless generator electrode, titration vessel kit, and an automated solvent exchange system for safer and more efficient workflows.
  • Innovative cell design simplifies cleaning and reduces cross-contamination risks.
  • Integrates seamlessly with balances for automatic, error-free sample weight transfer to enhance accuracy and efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Connector Types USB-A, RJ45, M8, M9, mini-DIN, 5-pin socket, 4-pin socket
Drift Dynamic
Languages Chinese, English, French, German, Italian, Korean, Polish, Portuguese, Russian, Spanish, Japanese, Thai, Turkish
Material PBT (Polybutylene Terephthalate), Stainless Steel (1.4301), Chrome-Plated ZnAl4Cu1, EPDM M Class (Ethylene Propylene Diene Monomer (M class) Rubber)
Type Titrator
Width (English) 5.3 in.
Depth (English) 7 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.