Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.716 M
Supplier:  Hach Company 2081801
Encompass
EDTA standard solution. For the determination of iron by Digital Titrator titration. 0.716 M. Digits = mg/L Fe in 50 mL
Catalog No. 50-199-5248