Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.0800 M
Supplier:  Hach Company 1436401
Encompass
For determination of Calcium and Total Hardness by Digital Titrator titration. 0.0800 M EDTA, Digits x 0.1 = mg/L CaCO3 in 100 mL sample. Approximately 40-100 tests
Catalog No. 50-199-5003