Learn More
CHEMetrics™ by AquaPhoenix Titrets DEHA Water Quality Visual Test Kit
SDP
Catalog No. 13299628
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-628 30
1 options
Catalog No. 13-299-628 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K3925
Add to Cart
Add to Cart
DEHA visual test kit utilizing Titrets titration cells with ceric sulfate titrant and ferroin indicator. Designed to measure DEHA in the 25 to 250 ppm range during lay-up.
  • Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, Activator Solution, titrettor, 25 mL sample cup, and instructions.
Specifications
Analytes DEHA
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 25 to 250 ppm
Type Water Quality Visual Test Kit