Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titriplex™ II, For Analysis, Ethylenedinitrilotetraacetic acid, ACS, MilliporeSigma™

Catalog No. M1084170250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1084170250 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1084170250 Supplier MilliporeSigma Supplier No. 1.08417.0250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color White
pH 2.5
Physical Form Fine Crystals or Crystalline Powder
Quantity 250 g
Assay Percent Range 99.4 - 100.6%
Grade ACS/Reagent Ph. Eur.
Ignition Point ≥200°C
Packaging Plastic bottle
Vapor Pressure ≤0.013 hPa (20°C)
Decomposition Information 220°C (Decomposition)
Density 0.86g/cm3 (20°C); Bulk Density: 600 kg/m3
Chemical Name or Material Ethylenedinitrilotetraacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H319: Causes serious eye irritation.
Hazard Statement P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 200-449-4
RTECSNumber AH4025000
storageNote1 Storage temperature: No restrictions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.