Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titriplex™ II, Ethylenedinitrilotetraacetic acid, ACS, MilliporeSigma™

Catalog No. M1084171000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1084171000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1084171000 Supplier MilliporeSigma Supplier No. 1084171000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ethylenedinitrilotetraacetic acid suitable for analytical applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Density 600kg/m3
pH 2.5
Ignition Point ≥200°C
Packaging Plastic Bottle
Quantity 1 kg
Solubility Information 0.5g/L
Vapor Pressure ≥0.013 hPa (20°C)
Formula Weight 292.24
Percent Purity ACS, Reag. Ph
Grade ACS
Physical Form Fine Crystalline to Crystalline Powder
Assay 99.4 - 100.6%
Chemical Name or Material Ethylenedinitrilotetraacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H319: Causes serious eye irritation.
Hazard Statement P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 200-449-4
RTECSNumber AH4025000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.