Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp Scientifica™ TITROLINE 5000 Automatic Titrator
SDP

Catalog No. 07222000
Change view
Click to view available options
Model:
Titroline Easy K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
07-222-000 Titroline Easy K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-222-000 Supplier Velp Scientifica™ Supplier No. R30800194
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Velp Scientifica™ TITROLINE 5000 Automatic Titrator with compact new titrator for simple routine titrations.

  • Titrations to pH, mV - endpoint (2 EP)
  • Titrations to one equivalence point (pH und mV) with dynamic or linear titration addition
  • Manual titrations with the controller
  • Dosing tasks
  • Number of methods: 3
  • Salt content in foodstuffs (cheese, soy sauce, ketchup)
  • Total acidity in wine and beverages
  • Nitrogen according to Kjeldahl
  • Chloride in dringing water and other samples
  • Alkalinity (p + m-value)
  • FOS/TAC

Specifications

Display Type 3.5 in. color touch screen
Type Automatic Titrator
For Use With (Application) Combined distillation and titration, for simultaneous Kjeldahl nitrogen/protein determination
Material Technopolymer
Model Titroline Easy K
Depth (Metric) 416 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.