Learn More
Medchemexpress LLC Tivanisiran (siRNA) | 1848224-71-4 | 97.5% | 12068.30 (AS: 5294.6 + SS: 6125.7) | C361H447N141O262P36 | 5 MG
SDP
Catalog No. 5000262963
Encompass_Preferred
Tivanisiran (SYL1001) is a double-stranded small interfering RNA (siRNA) designed to silence transient receptor potential vanilloid 1 (TRPV1) mRNA for research into dry eye disease. It is supplied as an oligonucleotide reagent intended for laboratory and preclinical gene-silencing studies.
  • Silences TRPV1 mRNA to reduce ocular discomfort in dry eye models.
  • Double-stranded siRNA sequence for targeted gene knockdown.
  • High purity suitable for research applications (97.5%).
  • Characterized molecular weight approximately 12,068 Da.
  • Available in small pack sizes suitable for experiments (for example, 5 mg).
  • Sequence information provided to support assay design and verification.
Catalog No. 50-002-62963 Supplier Medchemexpress LLC Supplier No. HY1325965MG
Tivanisiran (SYL1001) is a double-stranded small interfering RNA (siRNA) designed to silence transient receptor potential vanilloid 1 (TRPV1) mRNA for research into dry eye disease. It is supplied as an oligonucleotide reagent intended for laboratory and preclinical gene-silencing studies.
  • Silences TRPV1 mRNA to reduce ocular discomfort in dry eye models.
  • Double-stranded siRNA sequence for targeted gene knockdown.
  • High purity suitable for research applications (97.5%).
  • Characterized molecular weight approximately 12,068 Da.
  • Available in small pack sizes suitable for experiments (for example, 5 mg).
  • Sequence information provided to support assay design and verification.