Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tivanisiran (SYL1001) | 1848224-71-4 | 97.5% | 10MG
SDP

Catalog No. 5000261714
Encompass_Preferred

Tivanisiran (SYL1001) is a small interfering RNA (siRNA) developed to silence transient receptor potential vanilloid 1 (TRPV1) mRNA for preclinical research into dry eye disease. The material is supplied as a white to off-white solid for laboratory use only, with sequence, purity, solubility, and storage data provided by the supplier.

  • Designed to silence TRPV1 mRNA for ocular research.
  • Supplied as a white to off-white solid for laboratory use.
  • High purity, approximately 97.5%.
  • Sequence information for sense and antisense strands is provided.
  • Soluble in water up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored at -20°C as a solid; in solution, store at -80°C for long-term stability.

Catalog No. 50-002-61714 Supplier Medchemexpress LLC Supplier No. HY13259610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tivanisiran (SYL1001) is a small interfering RNA (siRNA) developed to silence transient receptor potential vanilloid 1 (TRPV1) mRNA for preclinical research into dry eye disease. The material is supplied as a white to off-white solid for laboratory use only, with sequence, purity, solubility, and storage data provided by the supplier.

  • Designed to silence TRPV1 mRNA for ocular research.
  • Supplied as a white to off-white solid for laboratory use.
  • High purity, approximately 97.5%.
  • Sequence information for sense and antisense strands is provided.
  • Soluble in water up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored at -20°C as a solid; in solution, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.